missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ IEX Selection Kit

Catalog No. 45002923
Change view
Click to view available options
Diameter (Metric):
7 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
45002923 7 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002923 Supplier Cytiva Supplier No. 28934388

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for use with syringe, peristaltic pump, or chromatography systems such as AKTA design for convenient media screening and optimization studies

  • Consists of five HiTrap columns, prepacked with Capto ion exchange chromatography media with different characteristics
  • For media screening; includes five 1mL HiTrap columns prepacked with Capto Q, Capto S, Capto DEAE, Capto MMC, Capto adhere, as well as connectors and instructions
  • For optimization studies; buffer composition, pH, flow rate, sample loading and elution schemes can be optimized with small sample quantities prior to scale-up
  • Capto Q is strong anion exchanger, Capto DEAE is weak anion exchanger, Capto S is strong cation exchanger
  • Capto MMC and Capto adhere media have multimodal characteristics that can be used to solve specific purification problems
  • Capto MMC is multimodal cation exchanger that is functional at high as well as at low ionic strength
  • Capto adhere is multimodal strong anion exchanger mainly designed for post-protein A purification of monoclonal antibodies
  • Prepacked columns and bulk media are available for larger scale preparative purifications

  • Max. pressure (over packed bed during operation): 3 bar (42psi)
  • Flow rate: <4mL/min.
  • Complete packsize: 5 x 1mL
  • Bed dimensions: 7 × 25mm
  • Bed volume: 1mL
BioProcess Media
  • pH Stability cleaning: 2 to 14
  • Matrix: highly cross-linked agarose with dextran surface extender
  • Average particle size: 90μm (d50V)
  • pH Stability working range: 2 to 12
  • Pressure/Flow specification: 0.3MPa at 700cm/hr., 1m diameter column, 20cm bed height
    
Column    
  • Column I.D.: 7mm
  • Material (column hardware): polypropylene (PP)

Specifications

Technique Ion Exchange Chromatography
Description HiTrap Capto IEX Selection Kit
Diameter (Metric) 7 mm
For Use With (Equipment) Syringe, peristaltic pump and other chromatography systems such as AKTAdesign