missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap Capto Butyl ImpRes

Catalog No. 45100700
Change view
Click to view available options
Bed Volume:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
45100700 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
The temperature difference between the fluid running through the column and the ambient temperature in the room should never be greater than 20°C
Catalog No. 45100700 Supplier Cytiva Supplier No. 17371911

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The temperature difference between the fluid running through the column and the ambient temperature in the room should never be greater than 20°C

A ready-to-use column prepacked with BioProcess Capto Butyl ImpRes resins for screening and small-scale protein purification using hydrophobic interaction chromatography.

  • Prepacked columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
  • High-throughput chromatography resin for intermediate and polishing steps in a downstream protein purification process.
  • Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide.
  • High binding capacity, high flow rate, and low back pressure reduces process cycle times and improves productivity at process scale.
  • Specifications

    Bed Volume 1 mL
    Flow Rate Up to 220 cm/h in a 1 m diameter column with a bed height of 20 cm at 20°C; measured using process buffers with the same viscosity as water at 300 kPa.
    For Use With (Application) Hydrophobic Interaction Chromatography 
    Height (Metric) 25 mm
    Ligand Type Butyl
    Max. Pressure 5 bar [0.5 MPa] (70 psi)
    Particle Size 40 μm
    pH Range pH 3 to 13
    Storage Requirements 20% ethanol at 4°C to 30°C