Learn More
Cytiva HiTrap™ Butyl-S FF Columns
Designed for purification of relatively strong hydrophilic molecules at low salt concentrations
$407.75 - $407.75
Specifications
Product Type HiTrap Butyl-S FF column
Ligand Type Butyl-S
Material Polypropylene
Particle Size 45 to 165 μm
pH Range 3 to 13
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Diameter (English) Inner Flow Rate Price Quantity  
Catalog Number Mfr. No. Quantity Diameter (English) Inner Flow Rate Price Quantity  
45-000-174
View Documents
Cytiva
17097813
5 x 1 mL 0.275 in. < 1 mL/min
Pack of 5 Each for $407.75
Only null left
Description
Description
  • Useful for removal of strong hydrophobic contaminants, such as lipoproteins, lipids, pigments and more from biological samples
  • Used successfully for large-scale purification of recombinant Hepatitis B virus surface antigen (rec-HBsAg) from CHO cells
  • Robust production with high reproducibility
Specifications
Specifications
HiTrap Butyl-S FF column
Polypropylene
3 to 13
Butyl-S
45 to 165 μm
6% Cross-linked Agarose