missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Histozyme

Catalog No. NBP350106
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP350106 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP350106 Supplier R&D Systems™ Supplier No. NBP350106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. For Enzyme pretreatment

Specifications

Content And Storage Store at 4°C in the dark. Do not freeze.
Product Line Novus
Quantity 15 mL
For Use With (Application) Immunohistochemistry-Paraffin