Learn More
Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassettes in Quickload™ Stacks (Taped)
Secure specimens during processing with Histosette™ II Tissue Processing/Embedding Cassettes in Quickload™ Stacks (Taped). Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
Supplier:  Simport™ Scientific M4925T
 View more versions of this product
Catalog No. 22038048
Only null left
Explore available promotions
Description
Description
  • In Quickload™ stacks
  • 45° anterior writing area
  • Open and close as often as necessary - cassettes will always relock securely
  • Back mounted tab allowing an unobstructed anterior marking area
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Specifications
2000/Pk.
Yellow
Acetal
Product Suggestions
Product Suggestions