missing translation for 'onlineSavingsMsg'
Learn More

HistoPrep™ Xylene, Fisherbrand™

Catalog No. 11002221
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002221 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002221 Supplier Statlab Supplier No. 11002221
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

Purified and filtered to deliver a high quality, consistent product.

  • Used as a clearing agent to make tissue transparent on microscope slides by removing paraffin wax from tissue sections, allowing stains to penetrate effectively, and ultimately making the tissue easier to examine under a microscope
  • Purified and filtered to deliver a high quality, consistent product
  • High solvency factor ensures quick and complete clearing
  • Free of impurities and water

Chemical Identifiers

CAS 1330-20-7

Specifications

Product Type Xylene
Quantity 1 gal.
Form Solution

Made in the USA