missing translation for 'onlineSavingsMsg'
Learn More

HistoPrep™ Methanol, Fisherbrand™

Catalog No. 11002212
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002212 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002212 Supplier Statlab Supplier No. 11002212

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

99% absolute methanol.

  • For general tissue processing as a substitute for ethanol, or as a cytology fixative.
  • Rapidly dehydrates and preserves cell morphology effectively

Chemical Identifiers

CAS 67-56-1

Specifications

Physical Form Aqueous Solution
Color Clear
Quantity 1 gal.
Percent Purity 100%
Grade ACS

Made in the USA