Learn More
Epredia™ Histoplast Paraffin
Catalog No. p-4532374
Change view
Click to view available options
Product Type:
Paraffin IM
Paraffin LP
Paraffin PE
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
8330 Paraffin PE
8331 Paraffin IM
8332 Paraffin LP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 8330 Supplier Epredia™ Supplier No. SH8330
Only null left
Meet the demanding and versatile needs of the laboratory with Epredia™ Histoplast Paraffin.
  • Made of paraffin and plastic polymers
  • Delivers excellent processing and yields thin, wrinkle-free sections
Ordering Information:
Packaged in convenient, easy-to-open, resealable 2 lb. bags. Also available in a premeasured paraffin kit for use on the Epredia™ Excelsior™ AS Tissue Processor (waste drawer included).
Specifications
Melting Point 56°C
Product Type Paraffin PE
Quantity 8 x 1 Kg