missing translation for 'onlineSavingsMsg'
Learn More

Histone, MP Biomedicals™

Catalog No. MP210072650
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210072650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210072650 Supplier MP Biomedicals Inc Supplier No. 0210072650
Only null left

Commonly used for chromosome-reconstitution studies. Histone, MP Biomedicals is an arginine-rich subgroup that is a suitable substrate for protein kinase C.

Chemical Identifiers

CAS 37244-51-2

Specifications

Quantity 50 mg
Physical Form Lyophilized Powder
Recommended Storage Store at 4°C.