missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Histone H3 Peptide (1-20)

Catalog No. PV5707
Encompass
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
PV5707 1 mL
1 options

Catalog No. PV5707

Supplier: Invitrogen™ PV5707

Only null left

Matched and validated lipid and peptide substrates are available to complement the Adapta™ Universal Kinase Assay.

Adapta™ Universal Kinase assays developed with these reagents allow for primary or secondary screening of kinase inhibitors.

Specifications

For Use With (Application) Kinase Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Pharma and Biopharma, Target and Lead Identification and Validation
Quantity 1 mL
Storage Requirements Substrates for Adapta™ Assays are shipped on dry ice and should be stored at -20°C.
Regulatory Status RUO - research use only
Structural Form ARTKQTARKSTGGKAPRKQLW
Form Liquid