Learn More
Azer Scientific Histology Tissue Control Slides
Designed for use in monitoring stain performance in the pathology laboratory
Supplier:  Azer Scientific ESCS130
 View more versions of this product
Catalog No. 22026724
Only null left
Includes: 10 unstained and 1 stained Unifrost slide
Description
Description
  • Prepared with paraffin-embedded human tissue sections
  • For QC of laboratory's staining processes
Specifications
Chemical Identifiers
Designed for use in monitoring stain performance in the pathology laboratory
10 unstained and 1 stained unifrost slide
11 Pack
Specifications
Helicobacter
Designed for use in monitoring stain performance in the pathology laboratory
10 unstained and 1 stained unifrost slide
11 Pack
Product Suggestions
Product Suggestions