missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Histamine, Free Base, ≥97%, Calbiochem™,
$201.00
Chemical Identifiers
| CAS | 51-45-6 |
|---|---|
| Molecular Formula | C5H9N3 |
| Molecular Weight (g/mol) | 111.15 |
| MDL Number | MFCD00005210,MFCD00128939 |
| InChI Key | NTYJJOPFIAHURM-UHFFFAOYSA-N |
| Synonym | histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine |
| PubChem CID | 774 |
| ChEBI | CHEBI:18295 |
| IUPAC Name | 2-(1H-imidazol-5-yl)ethan-1-amine |
| SMILES | NCCC1=CN=CN1 |
Chemical Identifiers
| C5H9N3 | |
| MFCD00005210,MFCD00128939 | |
| histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine | |
| CHEBI:18295 | |
| NCCC1=CN=CN1 |
Specifications
| 51-45-6 | |
| C5H9N3 | |
| histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine | |
| NCCC1=CN=CN1 | |
| 111.15 | |
| CHEBI:18295 | |
| Solid |
| White | |
| MFCD00005210,MFCD00128939 | |
| NTYJJOPFIAHURM-UHFFFAOYSA-N | |
| 2-(1H-imidazol-5-yl)ethan-1-amine | |
| 774 | |
| ≥97% by TLC |