missing translation for 'onlineSavingsMsg'
Learn More

Histamine Diphosphate Hydrate 98.0+%, TCI America™

Catalog No. p-7130372
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01471G 1 g
H01475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H01471G Supplier TCI America Supplier No. H01471G
Only null left

Chemical Identifiers

CAS 51-74-1
Molecular Formula C5H15N3O8P2
Molecular Weight (g/mol) 307.14
MDL Number MFCD00064316
InChI Key ZHIBQGJKHVBLJJ-UHFFFAOYSA-N
Synonym histamine acid phosphate, 2-1h-imidazol-4-yl ethanamine bis phosphate, histamine biphosphate, histamine dihydrogen phosphate, histamine phosphate 1:2, histamine phosphate, 1h-imidazole-4-ethanamine, phosphate 1:2, histamine phosphate usp, histamine positive, 4-2-aminoethyl imidazole bis dihydrogen phosphate
PubChem CID 65513
ChEBI CHEBI:51193
IUPAC Name 2-(1H-imidazol-5-yl)ethan-1-amine; bis(phosphoric acid)
SMILES OP(O)(O)=O.OP(O)(O)=O.NCCC1=CN=CN1

Specifications

Melting Point 133°C
Color White
Quantity 1 g
Formula Weight 307.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Histamine Diphosphate Hydrate
RTECSNumber NI5425000
TSCA Yes
Recommended Storage Refrigerator