missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine diphosphate, 98%
Description
Histamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Histamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Histamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 128°C to 132°C |
| Color | White |
| Quantity | 5 g |
| Sensitivity | Hygroscopic |
| Merck Index | 14,4719 |
| Solubility Information | Soluble in water. |
| IUPAC Name | 2-(1H-imidazol-5-yl)ethanamine;phosphoric acid |
| Formula Weight | 307.14 |
| Percent Purity | ≥99% |
| Physical Form | Powder |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H317-H318-H334-H335 |
| Hazard Statement | GHS H Statement H334-H318-H302-H335-H315-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c |
| EINECSNumber | 200-118-4 |
| RTECSNumber | NI5425000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only