missing translation for 'onlineSavingsMsg'
Learn More

Histamine diphosphate, 98%

Catalog No. AAJ6108806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6108806 5 g
AAJ6108814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6108806 Supplier Thermo Scientific Chemicals Supplier No. J6108806

Endogenous H1 and H2 agonist | CAS: 51-74-1 | C5H15N3O8P2 | 307.14 g/mol

Histamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histamine diphosphate is a potent agonist of histamine receptors. It activates nitric oxide synthetase and is a potent vasodilator.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 51-74-1
Molecular Formula C5H15N3O8P2
Molecular Weight (g/mol) 307.14
MDL Number MFCD00064316
InChI Key ZHIBQGJKHVBLJJ-UHFFFAOYSA-N
Synonym histamine acid phosphate, 2-1h-imidazol-4-yl ethanamine bis phosphate, histamine biphosphate, histamine dihydrogen phosphate, histamine phosphate 1:2, histamine phosphate, 1h-imidazole-4-ethanamine, phosphate 1:2, histamine phosphate usp, histamine positive, 4-2-aminoethyl imidazole bis dihydrogen phosphate
PubChem CID 65513
ChEBI CHEBI:51193
SMILES OP(O)(O)=O.OP(O)(O)=O.NCCC1=CN=CN1

Specifications

Melting Point 128°C to 132°C
Color White
Quantity 5 g
Sensitivity Hygroscopic
Merck Index 14,4719
Solubility Information Soluble in water.
IUPAC Name 2-(1H-imidazol-5-yl)ethanamine;phosphoric acid
Formula Weight 307.14
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Histamine diphosphate
Show More Show Less
Hazard Category H302-H315-H317-H318-H334-H335
Hazard Statement GHS H Statement
H334-H318-H302-H335-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
EINECSNumber 200-118-4
RTECSNumber NI5425000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only