missing translation for 'onlineSavingsMsg'
Learn More

Histamine dihydrochloride, 98+%

CAS: 56-92-8 | C5H11Cl2N3 | 184.06 g/mol

Supplier:  Thermo Scientific Chemicals L0919814

 View more versions of this product

Catalog No. AAL0919814


Only null left
Explore available promotions

Description

Description

Histamine dihydrochloride has been shown to activate nitric oxide synthetase and suppress or inhibit the generation of reactive oxygen species (ROS). Inhibition of ROS by histamine dihydrochloride allows activation of T cells and NK cells by IL-2. In a rat model, histamine dihydrochloride suppressed ROS generated by Kupffer cells through the H2 histamine receptor. It is a potent vasodilator and endogenous histamine receptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histamine dihydrochloride has been shown to activate nitric oxide synthetase and suppress or inhibit the generation of reactive oxygen species (ROS). Inhibition of ROS by histamine dihydrochloride allows activation of T cells and NK cells by IL-2. In a rat model, histamine dihydrochloride suppressed ROS generated by Kupffer cells through the H2 histamine receptor. It is a potent vasodilator and endogenous histamine receptor agonist.

Solubility
Very soluble in water and methanol. Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents. Store at 4°C.
Specifications

Specifications

Histamine dihydrochloride
245°C to 249°C
MFCD00012703
3624116
histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride
PPZMYIBUHIPZOS-UHFFFAOYSA-N
2-(1H-imidazol-5-yl)ethanamine;dihydrochloride
5818
≥98%
56-92-8
C5H11Cl2N3
25 g
14,4719
Very soluble in water and methanol. Soluble in ethanol.
[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
184.06
184.07

Chemical Identifiers

56-92-8
184.06
PPZMYIBUHIPZOS-UHFFFAOYSA-N
5818
[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
C5H11Cl2N3
MFCD00012703
histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride
2-(1H-imidazol-5-yl)ethanamine;dihydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c

H315-H317-H319-H334-H335

EINECSNumber : 200-298-4

RTECSNumber : MS1575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only