missing translation for 'onlineSavingsMsg'
Learn More

Histamine, 97%

CAS: 51-45-6 | C5H9N3 | 111.15 g/mol

$214.39 - $214.39

Chemical Identifiers

CAS 51-45-6
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD00005210,MFCD00128939
InChI Key NTYJJOPFIAHURM-UHFFFAOYSA-N
Synonym histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine
PubChem CID 774
ChEBI CHEBI:18295
IUPAC Name 2-(1H-imidazol-5-yl)ethanamine
SMILES NCCC1=CN=CN1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC411710010
SDS View Documents
Thermo Scientific Chemicals
411710010
1 g Glass bottle
Each for $214.39
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

51-45-6
111.15
NTYJJOPFIAHURM-UHFFFAOYSA-N
774
2-(1H-imidazol-5-yl)ethanamine
C5H9N3
MFCD00005210,MFCD00128939
histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine
CHEBI:18295
NCCC1=CN=CN1

Specifications

51-45-6
White to Yellow
Authentic
Glass bottle
MFCD00005210,MFCD00128939
15, 4756
NTYJJOPFIAHURM-UHFFFAOYSA-N
2-(1H-imidazol-5-yl)ethanamine
774
111.15
Crystalline Powder and/or Chunks
83°C to 84°C
167°C (0.8 mmHg)
96% min. (HPLC)
C5H9N3
1 g
histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine
NCCC1=CN=CN1
111.15
CHEBI:18295
97%
Histamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause resp

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 200-100-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only