missing translation for 'onlineSavingsMsg'
Learn More

Histamine, 97%

Catalog No. AC411710010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411710010 1 g Glass bottle
AC411710050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC411710010 Supplier Thermo Scientific Chemicals Supplier No. 411710010

CAS: 51-45-6 | C5H9N3 | 111.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-45-6
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD00005210,MFCD00128939
InChI Key NTYJJOPFIAHURM-UHFFFAOYSA-N
Synonym histamine, 1h-imidazole-4-ethanamine, 2-4-imidazolyl ethylamine, ergotidine, 2-1h-imidazol-5-yl ethanamine, ergamine, eramin, 5-imidazoleethylamine, theramine, free histamine
PubChem CID 774
ChEBI CHEBI:18295
IUPAC Name 2-(1H-imidazol-5-yl)ethanamine
SMILES NCCC1=CN=CN1

Specifications

Melting Point 83°C to 84°C
Color White to Yellow
Boiling Point 167°C (0.8 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 4756
Formula Weight 111.15
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Histamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause resp
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
EINECSNumber 200-100-6

RUO – Research Use Only