missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HisProbe™-HRP Conjugate

Catalog No. PI15165
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI15165 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI15165 Supplier Thermo Scientific™ Supplier No. 15165

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Directly detect histidine-tagged fusion proteins and other histidine-rich proteins with this nickel-activated horseradish peroxidase (HRP) derivative.

Thermo Scientific™ HisProbe-HRP is an nickel (Ni2+)-activated derivative of horseradish peroxidase (HRP). This product has been optimized for direct detection of recombinant histidine-tagged proteins and other histidine-rich proteins.

Unlike anti-6xHis antibodies, HisProbe-HRP can recognize polyhistidine tags independent of adjacent tags. The detection kit includes blocking buffers and chemiluminescent substrate for sensitive Western blot detection via the HisProbe-HRP Conjugate. The active ligand in HisProbe-HRP is a tridentate chelator that allows Ni2+ to be bound in active form for subsequent interaction and detection fo target molecules. The active chelator has binding capabilities similar to iminodiacetic acid (IDA) when used for immobilized metal affinity chromatography (IMAC).

Highlights:

  • Fast – His-tag detection is achieved with a one-step probe incubation that eliminates the need to run a lengthy two-step primary/secondary antibody reaction protocol

  • Economical – no need to purchase expensive primary/secondary antibodies; considerably less costly than anti-6xHis-tag antibodies

  • High specificity – HisProbe-HRP can recognize his-tags independent of adjacent tags

  • Less steric hindrance – HisProbe-HRP is smaller than antibodies and detects his tagged fusion proteins that are often undetectable when using some anti-his-tag antibodies

Specifications

Description Pierce HisProbe-HRP Conjugate
Product Type HRP Conjugate
Detection Method Chemiluminescence
Form Solid
Product Line HisProbe
Quantity 2 mg
Reactivity All species
Content And Storage Storage: Upon receipt store product at 4°C. Product is shipped at ambient temperature.

For Research Use Only. Not for use in diagnostic procedures.