missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns
Description
- Ready-to-use columns for convenient process development
- Rigid, highly cross-linked agarose matrix
- Bed height of 10cm is designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20cm bed height
- Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
MabSelect
- For capture of MAbs from large sample volumes
- Prepacked with protein A-derived media
- Compatible with high flow rates and high pressure when scaling up
- Increased binding capacity enables purification of MAbs from high-level expression feedstock
- Also optimized for Fc-fusion proteins
- Very low, unspecific binding due to high ligand selectivity and matrix hydrophilicity
- Prepacked with protein A-derived media
- With novel, alkali-tolerant recombinant protein A ligand that provides greater stability and allows use of 0.1 to 0.5M sodium hydroxide for cleaning-in-place (CIP)
- Enhanced protease resistance resulting in lower ligand leakage
- High dynamic binding capacity
Specifications
Specifications
| Buffer | 8M Urea, 6M Guanidine Hydrochloride, 2% Benzyl Alcohol or 20% Ethanol |
| Column Type | HiScreen MabSelect Prepacked Column |
| Includes | Stoppers at the inlet and outlet |
| Kit Size | 1 x 4.7 mL |
| Bed Support | Filter |
| Diameter (English) Inner | 0.3 in. |
| Binding Capacity | ∼30 mg Human IgG |
| Diameter (Metric) Inner | 7.7 mm |
| Product Type | HiScreen MabSelect Prepacked Column |
| Height (English) Bed | 3.93 in. |
| Show More |