missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Capto™ MMC ImpRes Multimodal Chromatography Column

Catalog No. 45002503
Change view
Click to view available options
Particle Size:
40 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size
45002503 40 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002503 Supplier Cytiva Supplier No. 17371620

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use column for method optimization and parameter screening

  • Prepacked with different BioProcess™ media for convenient process development
  • Bed height: 10cm
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume for fast results and minimal sample/buffer consumption
  • Displays broad pH/salt operational window
  • Enables use of platform approach to MAb process development
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • High-resolution beads and ligand selectivity ensures high yield

  • Reproducible results
  • Allows effective, high-resolution polishing of MAb
  • Binds proteins at high or low ionic strength
  • Reduces sample cost and buffer consumption

Avoid using oxidizing agents, cationic detergents.

Specifications

For Use With (Application) HPLC, Parameter Screening, Optimization Method
Max. Pressure 300,000 psi
Particle Size 40 μm