missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Capto™ Adhere Impres Multimodal Chromatography Column
GREENER_CHOICE

Catalog No. 45002501
Change view
Click to view available options
Particle Size:
40 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size
45002501 40 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002501 Supplier Cytiva Supplier No. 17371520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Certificate of Analysis

Ready-to-use column for method optimization and parameter screening

  • Prepacked with different BioProcess™ media for convenient process development
  • Bed height: 10cm
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume for fast results and minimal sample/buffer consumption
  • Reduces sample cost and buffer consumption
  • Binds proteins at high or low ionic strength
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • High-resolution beads and ligand selectivity ensures high yield

  • Reproducible results
  • Allows effective, high-resolution polishing of MAb

Avoid using oxidizing agents, anionic detergents.

Specifications

Certifications/Compliance Certificate of analysis
For Use With (Application) MAbs Purification
Ion Exchange Group Anion
Max. Pressure 43.51 psi
Particle Size 40 μm
Type Chromatography Column
Film Thickness 40 μm
Material Polypropylene
Packing Material HiScreen
Diameter (Metric) Inner 7.7 mm