missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ HiPur™ 10% Neutral Buffered Formalin
Specifications
Specifications
| Color | Colorless |
| Content And Storage | Room temperature |
| Concentration | 0.1 |
| Quantity | 5 gal. |
| For Use With (Application) | Histological fixative to fix tissues |