missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ HiPur™ 10% Neutral Buffered Formalin

Catalog No. 22110873
Change view
Click to view available options
Quantity:
5 gal.
4 x 1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22110873 4 x 1 gal.
22110875 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22110873 Supplier Epredia™ Supplier No. 94001
Only null left

Choose the option that best fits your workflow, HiPur™ 10% neutral buffered formalin is available in one and five gallon sizes.

Specifications

Color Colorless
Content And Storage Room temperature
Concentration 0.1
Quantity 4 x 1 gal.
For Use With (Application) Histological fixative to fix tissues