missing translation for 'onlineSavingsMsg'
Learn More

CHECKERS™ Hinged 5-Channel Cable Protector End Cap

Catalog No. 19085965
Change view
Click to view available options
Color:
Orange
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19085965 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19085965 Supplier CHECKERS™ Supplier No. GDEC5X125O

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cable protector end cap is lightweight and easy to transport, set up, disassemble, and store. All-weather polyurethane construction with dog bone connection style

  • Dog bone connection style
  • 5 channels
  • Channel width: 1.25 in.
  • Channel height: 1.25 in.
  • For use with vehicle and pedestrian traffic
  • Ideal for amusement parks, entertainment venues, sporting events, industrial, construction, commercial applications

Specifications

Material Polyurethane
For Use With (Equipment) Cable protector
Height (English) 2 in.
Packaging Quantity 1 Ea.
Width (English) 19.75 in.
Color Orange