missing translation for 'onlineSavingsMsg'
Learn More

Hinge Lifters

Catalog No. S99536
Change view
Click to view available options
Dimensions (L x W):
2 x 2 in. (5.1 x 5.1 cm)
2 x 4 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S99536 2 x 4 in.
S69016A 2 x 2 in. (5.1 x 5.1 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S99536 Supplier Customer Channels Group (Ccg) Supplier No. 131LT

missing translation for 'validMainframeMsg'

Sized for multiple fingerprints or full palm prints.

By hinging the two together, you can lift, seal and protect fragile latent prints in one easy step.

  • Pre-assembled devices combine 5mil clear acetate-coated lifting medium with acrylic adhesive and 10mil backing sheet
  • Immediate comparisons may be made with suspect prints or used directly for contact prints or enlargements
  • Transparent

Specifications

Dimensions (L x W) 2 x 4 in.
Product Type Hinge Lifter
For Use With (Application) For multiple fingerprints or partial palm prints
Color Clear