missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiLoad™ 16/600 Superdex™ 30 pg Prepacked Column

Catalog No. 45002486
Change view
Click to view available options
Diameter (Metric):
16 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
45002486 16 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002486 Supplier Cytiva Supplier No. 28989331

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Dismantling tool, support screen, net ring (10μm), O-ring, stop plugs, and instructions.

Designed for high-resolution preparative gel filtration chromatography separating peptides

  • Prepacked XK columns
  • 1/16 in. fittings for convenient connections to KTA design systems
  • High resolution with short run times and good recovery
  • High capacity at high flow rates
  • Separations can be scaled up to production levels
  • Prepacked for convenience and reproducibility
  • Produced by covalent bonding of dextran to highly cross-linked agarose
  • Separation properties of these media are determined by the dextran component
  • BioProcess medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions

  • Maximum pressure: 42psi
  • Bed dimensions: 16 × 600mm
  • Flow rate: <1.0mL/min.
  • Bed volume: 120mL
  • Flow velocity: 10 to 50cm/hr.
  • Sample volume: 5mL
  • Theoretical plates: >13000m-1
  • Complete pack size: 1 × 120mL
Media
  • pH Stability working range: 3 to 12
  • pH Stability cleaning: 1 to 14
  • Average particle size: 34μm
  • Matrix: composite of cross-linked agarose and dextran
Column
  • Material (column tube): borosilicate glass
  • Porosity: 10μm
  • Bed support: net
  • Material (Bed support): polypropylene (PP) (reinforced glass fiber), polyamide (PA)
  • Material (Column hardware): borosilicate glass, polyamide, fluororubber, ethylene propylene
  • diene (EPDM), ETFE, superpolyoxymethylene
  • Outlet I.D.: 0.8mm (0.031 in.)
  • Column I.D.: 16mm (0.62 in.)
  • Inlet I.D.. 0.8mm (0.031 in.)

Specifications

Description Superdex 30 PG size exclusion chromatography media prepacked into HiLoad 26/600 column
Diameter (Metric) 16 mm