Learn More
Pyramex™ Highlander Plus™ Safety Eyewear
Fully adjustable elastic strap with quick release attaches to temples providing a secure fit
Supplier: Pyramex™ SBG5080DT
Description
Specifications
| Anti-fog | |
| Indoor/Outdoor, Mirror | |
| Black | |
| Nylon |
| ANSI Z87.1, CSA Z94.3-07 | |
| Ballistic Safety Glasses | |
| Polycarbonate |