Learn More
Synthware™ High Volume Double Oblique Bore Glass Stopcock with Constant Pressure Retention
Specifications
Specifications
| Type | Retainer Click |
| Plug Bore | 4 mm |
| Material | Borosilicate Glass |
| For Use With (Application) | Organic Synthesis |
| For Use With (Equipment) | 16.2/56 Glass Stopcock |
| Includes | Constant pressure retention spring and clip |
| Plug Material | Glass |
| Connectors | Ground Glass |