missing translation for 'onlineSavingsMsg'
Learn More

Fischer Technical Company™ High Vacuum Tubing and Clamp Set

Catalog No. S04551
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S04551 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04551 Supplier Fischer Technical Company™ Supplier No. HVGTUBING/SET

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High vacuum tubing can withstand 25 μm of pressure.

  • Inner Diameter: 0.5 in. (1.3 cm)
  • Ships complete with two hose clamps
  • Tubing is 5 ft. (18.3 m) in length

Specifications

Grade 6 to 12
Format Kit
Product Type High Vacuum Pump Tubing
Subject Science