missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ High Resolution Melt Software v3.0.1

Catalog No. 4461357
Encompass
Change view
Click to view available options
Quantity:
1 License
10 Licenses
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4461357 1 License
4461456 10 Licenses
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4461357 Supplier Applied Biosystems™ Supplier No. 4461357

High Resolution Melt Software v3.0.1 is an all-new and improved tool for High Resolution Melting (HRM) analysis.

HRM analysis is a post-PCR analysis method used to identify variation in nucleic acid sequences. The method is based on detecting small differences in PCR melting (dissociation) curves. It is enabled by high-brightness, dsDNA-binding dyes used in conjunction with real-time PCR instrumentation that has precise temperature ramp control, advanced data capture capabilities, and access to software designed specifically for HRM analysis. High Resolution Melt Software v3.0.1 brings HRM to the StepOne™ and StepOnePlus™ Real-Time PCR Systems, and the 7500 Fast Real-Time PCR System, and contains features that make fast and accurate HRM results more accessible than ever.

Key product features:

• Minimal user input to achieve results—software automatically applies default/saved analysis settings upon opening an experiment file
• Easy recall—new Analysis Settings Library saves settings for future experiments
• Greater accuracy—clustering algorithm accurately distinguishes control and variant genotypes
• Increased sensitivity—software allows input of expected number of clusters for difficult-to-detect SNPs (class 4)
• Ability to achieve more in one run—separate analysis of multiple assays run on a single plate
• Shortcuts in experiment setup—direct copy and paste functions with Excel; well information and plate layout clearly displayed in Plate View

Minimize User Input to Achieve Results and Easily Recall Analysis Settings
High Resolution Melt Software v3.0.1 automatically applies default or saved analysis settings upon opening an experiment file, which allows the user to achieve results with minimal input. It also includes a new Analysis Settings Library, allowing the user to save optimized settings and easily recall them for use in future experiments.

Realize Greater Accuracy and Increase Your Sensitivity
The v3.0.1 software utilizes an improved clustering algorithm that helps to accurately distinguish control and variant genotypes, and allows the user to input the expected number of clusters for increased sensitivity for difficult SNP genotyping.

Achieve More in One Run
With the v3.0.1 software, you now have the ability to conduct separate analyses of multiple assays run on a single plate.

Get Your Experiments Up and Running Fast with Shortcuts in Setup
Users of High Resolution Melt Software v3.0.1 can copy and paste directly from Excel spreadsheets to set up experiments quickly. Plate View also clearly displays well information and the plate layout to ease plate setup.

Specifications

Content And Storage Software CD, License Key
For Use With (Equipment) 7500 Fast System, StepOne™, StepOnePlus™
Software Type HRM Analysis Software
Product Type Software
For Use With (Application) Real Time PCR (qPCR)
Operating System Windows 7, Windows XP Pro SP2
Quantity 1 License
How long should the amplicon be for HRM analysis?

We recommend to use 50-250 bp long PCR amplicons. Typically, shorter amplicons can distinguish the genotypes for a SNP better, especially for Type III and Type IV SNPs. This is simply because a single base variation affects the melting behavior of a 100 bp amplicon stronger than of a 500 bp amplicon, for example. In longer fragments, the risk of covering multiple mutations is also increased.

What can be used as positive control reactions in HRM analysis?

Positive control reactions should contain template DNA with a known sequence. In SNP genotyping experiments, this could be a sample with a known genotype. Positive control(s) for all genotypes should be included where possible to serve as a reference in melting curve comparison and assigning genotypes for test samples. In mutation scanning experiments, a sample with a wild type sequence could serve as a positive control. The controls should preferably have the same DNA concentration as their corresponding test samples. Control DNA should also be eluted and/or diluted in the same buffer as the samples.

In HRM analysis, when is it better to use a 3-step cycling protocol rather than a 2-step protocol?

A 3-step cycling protocol is recommended for the analysis of complicated (especially Type IV SNP) targets, amplicons longer than 200 bp, and amplicons with a primer annealing temperature that is less than 60 degrees C.

The HRM software will not accept my calibration file. What is wrong?

Check that there are no outliers on the plate. You cannot omit any wells on the HRM calibration plate.

What instruments are compatible with High Resolution Melt (HRM)?

HRM can be performed on the 7500 Fast, 7900HT Fast, StepOne/StepOnePlus instruments and requires a separate purchase of HRM software. The following instruments are also compatible with HRM and have a built-in analysis module within the instrument software: ViiA7, QuantStudio 6 and 7 Pro, QuantStudio 6 and 7 Flex, QuantStudio 12K Flex, QuantStudio 1, QuantStudio 3, and QuantStudio 5. To activate the HRM module, you will need to purchase a license.

What reagents do I need to perform an HRM experiment?

To perform an HRM experiment, you will need the reagents to calibrate your instrument for the HRM dye and the software to perform the analysis.

What can I do with HRM?

High resolution melt (HRM) can be used in genotyping, mutation scanning, and methylation studies.

What are the license renewal requirements for the High Resolution Melt (HRM) software?

If you are using the QuantStudio 3/QuantStudio 5 Real-Time PCR Systems, 7500 Real-Time PCR System (with SDS v2.0.5 or later), or StepOne/StepOnePlus Real Time PCR Systems, you can purchase High Resolution Melt (HRM) Software v3.1 (https://www.thermofisher.com/order/catalog/product/A30150?ICID=search-product). Once activated, this software will not require annual renewal. If you have the older HRM software v3.0.1 (for use with StepOne Plus, StepOne or the 7500 Fast (SDS v2.0.5 or later) Real-Time PCR System, it will require license renewal on an annual basis. For use on the 7900HT Fast Real-Time PCR System, please use HRM Software v2.0.1 (https://www.thermofisher.com/order/catalog/product/4397808?ICID=search-product). This software will require license renewal on an annual basis. When using either the ViiA 7, QuantStudio 6/QuantStudio 7 or QuantStudio 12K Flex Real-Time PCR Systems, the HRM software is already included in the instrument's application software, but requires activation of the HRM module within the instrument software by purchase of the license. This license requires an annual renewal.