missing translation for 'onlineSavingsMsg'
Learn More

Cytiva High Purity rNTPs: Solid

Catalog No. 45001323
Change view
Click to view available options
Format:
Solid
Product Type:
ATP, disodium, crystalline
CTP, sodium, crystalline
GTP, sodium, crystalline
Quantity:
1 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Product Type Quantity
45001323 Solid ATP, disodium, crystalline 25 g
45001324 Solid CTP, sodium, crystalline 1 g
45001337 Solid GTP, sodium, crystalline 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45001323 Supplier Cytiva Supplier No. 27100603
Only null left

Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity

  • Each lot is tested for ribonuclease contamination to ensure success in your application
  • High purity: >98% triphosphate

Specifications

Format Solid
Description Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
pH 7.5
Molecular Weight (g/mol) 551.15
Product Type ATP, disodium, crystalline
Purity ≥98% triphosphate
Quantity 25 g
Color White
For Use With (Application) in-vitro transcription, amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling