missing translation for 'onlineSavingsMsg'
Learn More

Cytiva High Purity rNTP Solutions

Use for in vitro transcription or for reactions requiring rNTPs of high purity. Cytiva Lifescience™ High Purity rNTP Solutions provide easy-to-use, time-saving solutions.

$117.97 - $117.97

Specifications

Format Liquid
Description Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
pH 7.5
Concentration 100 mM
Purity ≥98% triphosphate
View More Specs

Products 4
Catalog Number Mfr. No. Format Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Format Product Type Quantity Price Quantity  
45-001-343
View Documents
Cytiva
27206601
Liquid CTP, 100 mM solution 25 μmol
Each for $117.97
Only null left
 
45-001-345
View Documents
Cytiva
27207601
Liquid GTP, 100 mM solution 25 μmol
Each for $117.97
Only null left
 
45-001-347
View Documents
Cytiva
27208601
Liquid UTP, 100 mM solution 25 μmol
Each for $117.97
Only null left
 
45-001-341
View Documents
Cytiva
27205601
Liquid ATP, 100 mM solution 25 μmol
Each for $117.97
Only null left
 
Description

Description

  • Each lot is tested for ribonuclease contamination to ensure success in your application
  • High purity: >98% triphosphate

Specifications

Specifications

Liquid
7.5
≥98% triphosphate
Colorless
Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
100 mM
25 μmol
in-vitro transcription, amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'