Learn More
Idex High-Pressure, Zero Dead Volume Unions
PEEK ZDV unions provide accurate results
Supplier: Idex UP P 704/DUP
Description
- Inert PEEK fluid path
- 10-32 coned configuration
Chemical Identifiers
| Union | |
| 6000 psi | |
| PEEK |
Specifications
| Union | |
| 6000 psi | |
| PEEK |
| 0.02 in. | |
| 0.283 μL | |
| 1/16 in. O.D. Tubing |