missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ High-Power USB Wi-Fi Module

Catalog No. A26774
Encompass
Change view
Click to view available options
Quantity:
1 module
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26774 1 module
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26774 Supplier Applied Biosystems™ Supplier No. A26774
Save Up to $945 with a Cell Counter Starter Kit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-Power USB Wi-Fi Module

Upgrade your wireless-enabled Applied Biosystems™ instrument with this High-Power USB Wi-Fi Module for high speed transfer rates and maximum range of connectivity during remote monitoring of your instrument.

Key features and benefits include:
  • Encrypted data security standards
  • Leading wireless technology standard for top performance and range
  • Maximum speed (up to 300 Mbps) for fast wireless transfer rates
  • Maximum range with built-in high-power antenna

    Note: This module is not suitable for the QuantStudio™ 3D Digital PCR Instrument. A different Wi-Fi module is offered for this instrument (A28598).
  • Specifications

    For Use With (Equipment) End-point Thermal Cycler
    Product Line Thermal Cycler Accessories
    Product Type USB Wi-Fi Module
    For Use With (Application) Real Time PCR (qPCR)
    Shipping Condition Room Temperature
    Quantity 1 module

    For Research Use Only. Not for use in diagnostic procedures.