Learn More
Thermo Scientific™ High Performance Microtube with Snap-on Seal Closure
Catalog No. 314352h01
Change view
Click to view available options
S110-AT, S100-AT4, S100-AT5, S100-AT6, and S80-AT3 rotors with adapters. S55-A2 and S45-A rotor are used without adapters.
Catalog No.
314352H01 S110-AT, S100-AT4, S100-AT5, S100-AT6, and S80-AT3 rotors with adapters. S55-A2 and S45-A rotor are used without adapters.
1 options
Catalog No. 314352H01 Supplier Thermo Scientific™ Supplier No. 314352H01
Recommended when using microcentrifuge tubes in Thermo Scientific micro-ultracentrifuge rotors.
  • 1.5 mL High Performance Microtube with Snap-on Seal Closure.
  • Recommended when using microcentrifuge tubes in Thermo Scientific micro-ultracentrifuge rotors.
  • The maximum RCF of this microtube is 201,000 x g
Specifications
For Use With (Equipment) S110-AT, S100-AT4, S100-AT5, S100-AT6, and S80-AT3 Rotors with Adapters. S55-A2 and S45-A Rotor Are Used Without Adapters.
Capacity 1.5 mL
Max. RCF 201,000 x g
Max. Speed 55,000 rpm
Diameter (Metric) Outside 11 mm
Length (Metric) 42 mm
Quantity 100/Pk.
Product Type Microcentrifuge Tube