missing translation for 'onlineSavingsMsg'
Learn More

CoorsTek™ High-Form Alumina Crucibles

Withstand temperatures up to 1750°C (3182°F)

Supplier:  CoorsTek™ CRS65501

 View more versions of this product

Catalog No. 12-220A


Only null left

Description

Description

  • High-purity (99.8%) recrystallized aluminum oxide
  • Nonporous, gas and vacuum tight
  • Can be used in oxidizing and reducing atmospheres and with all kinds of metals
  • Covers sold separately

Specifications

Specifications

Alumina
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions