Learn More
StatLab™ High Definition Acid Rinse 3.5% Solution
Description
- Premixed, safe and ready to use
- Provides crisp nuclear chromatin detail and eliminates any undesired background staining when used with StatLab™ Reserve and Vintage Hematoxylins
Specifications
Specifications
| Concentration | 3.5% |
| Quantity | 4 x 1 gal. |
| Product Type | Acid Rinse |