missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HIC (Hydrophobic Interaction Chromatography) Media

Catalog No. 45002995
Change view
Click to view available options
Product Type:
Capto Phenyl (High Sub) media
Ligand Type:
Phenyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Ligand Type
45002995 Capto Phenyl (High Sub) media Phenyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002995 Supplier Cytiva Supplier No. 17545101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Features high chemical stability, properties to increase productivity and epoxy coupling

  • High flow rate, high capacity and low backpressure reduce cycle times
  • Chemically stable to commonly used aqueous buffers
  • Epoxy chemical coupling

  • Capto Phenyl (high sub) has a hydrophobic ligand of phenyl and is a hydrophobic interaction chromatography medium used for protein purification
  • Capto Butyl has a hydrophobic ligand of butly and a highly cross-linked agarose matrix

Used in capture and intermediate stages of protein purification

Specifications

Product Type Capto Phenyl (High Sub) media
Flow Rate 600 cm/Hoμr
For Use With (Application) For Used in The Capture and Intermediate Stages of Protein PurifiCatIon
Quantity 25 mL
Ligand Type Phenyl
Media Type Highly Cross-Linked Agarose
Max. Pressure 300 kPa
Particle Size 75 μm
pH Range 3 to 13
Storage Requirements 4°C to 30°C
Storage Buffer 20% Ethanol
Binding Capacity 27 mg BSA/mL
Height (Metric) Bed 20 cm
Show More Show Less