Learn More
Brainbits HIBERNATE E LOW FLUORESCENCE
Catalog No. NC0285514
Encompass
HIBERNATE E LOW FLUORESCENCE, 500ML Hibernate is a nutrient medium for the maintenance of neural tissue or cells in ambient carbon dioxide levels (US Patent 6,180,404). Hibernate-E is useful for embryonic tissue.
Catalog No. NC0285514 Supplier Brainbits Supplier No. HELF
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
HIBERNATE E LOW FLUORESCENCE, 500ML Hibernate is a nutrient medium for the maintenance of neural tissue or cells in ambient carbon dioxide levels (US Patent 6,180,404). Hibernate-E is useful for embryonic tissue.