missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One HiBase Solid-Bottom 1536-well Microplate

Free of detectable DNase, RNase, human DNA and pyrogens

Supplier:  Greiner Bio-One 782101

Catalog No. 07-000-003

Only null left
Explore available promotions

Description

Description

Alphanumeric well coding
Specifications

Specifications

Free of detectable DNase/RNase, human DNA and pyrogens
Without Lid
32 x 48
Clear
1536
Non-sterile
Rounded Square
1800 x g (swinging-bucket rotor)
15
-20°C to +60°C
Standard
2.3 mm 2
Not Autoclavable
Polystyrene
3 to 10 μL
Non-treated
Yes
60
Yes
128 x 85 x 10 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions