missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyramex Safety Products Hi-Vis Safety Vest with Contrasting Reflective Tape
Pyramex Safety™ Hi-Vis Safety Vest with Contrasting Reflective Tape features a 4 inch. contrasting color trim around bottom and over shoulders which form an X on the back.
Supplier: Pyramex Safety Products RCZ2410XL
Specifications
| Zipper | |
| X-Large | |
| 2 mic holder loops | |
| 2 in. silver reflective tape, 4 in. contrasting color trim |
| Lime | |
| Solid Polyester Front, Mesh Back | |
| 8: 5 outside, 3 inside |