missing translation for 'onlineSavingsMsg'
Learn More

HF Scientific™ ProCal™ Turbidity Standard Reagents Kit for Hach™ 2100AN Turbidimeter

Standards not harmful to the environment or to worker safety

$392.04

Specifications

For Use With (Equipment) Hach 2100AN Turbidimeter
Certifications/Compliance EPA
View More Specs

Products 1
Catalog Number Mfr. No. Type Volume (Metric) Turbidity Price Quantity & Availability  
Catalog Number Mfr. No. Type Volume (Metric) Turbidity Price Quantity & Availability  
15393616
Encompass
View Documents Promotion Details
HF Scientific™
HF 52300
Primary Turbidity Standard 250 mL 20 NTU
Each for $392.04
Only null left
 
Description

Description

  • Pre-diluted
  • Do not include formazin
  • Preservative used enhances stability
  • Biodegradable, safe and non-toxic
  • Ready and easy to use
  • Factory certified by HF scientific
  • Custom NTU produced
  • Available in different size bottles

Specifications

Specifications

Hach 2100AN Turbidimeter
EPA
Videos
Safety and Handling

Safety and Handling

ShelfLife : 2 Years

SDS
Documents

Documents