Learn More
HF Scientific™ Powder Pop™ DPD Reagent for Chlorine Testing DFS Item
For testing free or total chlorine
Supplier:  HF Scientific™ 10315
 View more versions of this product
Catalog No. 15-393-67
Only null left
Description
Description
  • Deliver doses of DPD reagent measured for 5, 10 or 25mL (0.2, 0.3 or 0.8 oz.) testing samples
  • Individually packaged
Specifications
Specifications
25 mL
200/Pk.
0.84 oz.
For testing total chlorine
DPD Reagent
Product Suggestions
Product Suggestions