Learn More
HF Scientific™ Chlorine Micro Check Test Strips
Detect free or total chlorine concentrations as low as 0.05ppm
Supplier: HF Scientific™ HF 09941
Includes: 50 test strips are supplied in bottle labeled with instructions, color-comparison scale, and expiration date
Description
Detect free or total chlorine concentrations as low as 0.05ppm
Test strips for field testing and quality-control procedures; provide a safe and economical alternative to reagent chlorine testing.
- Accurate results produced in less than one minute
- Easy to use—simply dip strip into sample or hold under liquid stream to saturate, remove, and match color to the color-comparison scale
Chemical Identifiers
| Strip | |
| 50 test strips are supplied in bottle labeled with instructions, color comparison scale, and expiration date | |
| >1 min. |
| For detection of total chlorine, field testing and quality-control procedures, provide a safe and economical alternative to reagent chlorine testing | |
| 50 Pack |
Specifications
| Strip | |
| 50 test strips are supplied in bottle labeled with instructions, color comparison scale, and expiration date | |
| High range: 0.0, 0.1, 0.2, 0.5, 0.8, 4.0, 10.0; Low range: 0.0, 0.05, 0.1, 0.15, 0.2, 0.5, 1.0 | |
| Total Chlorine Test Strip |
| For detection of total chlorine, field testing and quality-control procedures, provide a safe and economical alternative to reagent chlorine testing | |
| 50 Pack | |
| >1 min. |