missing translation for 'onlineSavingsMsg'
Learn More

Hexylphosphonic Acid 98.0+%, TCI America™

Catalog No. H15511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15511G 1 g
H15515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15511G Supplier TCI America Supplier No. H15511G

Chemical Identifiers

CAS 4721-24-8
Molecular Formula C6H15O3P
Molecular Weight (g/mol) 166.157
MDL Number MFCD00013961
InChI Key GJWAEWLHSDGBGG-UHFFFAOYSA-N
Synonym n-hexylphosphonic acid, phosphonic acid, p-hexyl, hexylphosphonicacid, p-hexyl-phosphonic acid, hexylphosphonic acid
PubChem CID 312552
IUPAC Name hexylphosphonic acid
SMILES CCCCCCP(=O)(O)O

Specifications

Melting Point 108°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 166.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hexylphosphonic Acid
EINECSNumber (2)-1995
TSCA No