missing translation for 'onlineSavingsMsg'
Learn More

Hexylmagnesium chloride, 2M solution in THF, AcroSeal™

CAS: 44767-62-6 | C6H13ClMg | 144.93 g/mol

$122.50 - $122.50

Chemical Identifiers

CAS 44767-62-6
Molecular Formula C6H13ClMg
Molecular Weight (g/mol) 144.93
MDL Number MFCD06798087
InChI Key ODOOOEXDBNGOOF-UHFFFAOYSA-M
Synonym hexylmagnesium chloride, 1-hexylmagnesium chloride, hexylmagnesium chloride solution, hexyl magnesium chloride, n-hexylmagnesium chloride, chloro hexyl magnesium, odoooexdbngoof-uhfffaoysa-m, 1-hexylmagnesium chloride, 1m in methf, hexylmagnesium chloride2.0m solution, hexylmagnesium chloride solution, 2.0 m in thf
PubChem CID 10877247
SMILES CCCCCC[Mg]Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434681000
SDS View Documents
Thermo Scientific Chemicals
434681000
100 mL AcroSeal™ Glass Bottle
Each for $122.50
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

44767-62-6
144.93
ODOOOEXDBNGOOF-UHFFFAOYSA-M
10877247
C6H13ClMg
MFCD06798087
hexylmagnesium chloride, 1-hexylmagnesium chloride, hexylmagnesium chloride solution, hexyl magnesium chloride, n-hexylmagnesium chloride, chloro hexyl magnesium, odoooexdbngoof-uhfffaoysa-m, 1-hexylmagnesium chloride, 1m in methf, hexylmagnesium chloride2.0m solution, hexylmagnesium chloride solution, 2.0 m in thf
CCCCCC[Mg]Cl

Specifications

65.0°C to 68.0°C
Brown to Brown-Gray
100 mL
MFCD06798087
hexylmagnesium chloride, 1-hexylmagnesium chloride, hexylmagnesium chloride solution, hexyl magnesium chloride, n-hexylmagnesium chloride, chloro hexyl magnesium, odoooexdbngoof-uhfffaoysa-m, 1-hexylmagnesium chloride, 1m in methf, hexylmagnesium chloride2.0m solution, hexylmagnesium chloride solution, 2.0 m in thf
ODOOOEXDBNGOOF-UHFFFAOYSA-M
chloro(hexyl)magnesium
10877247
1.90 to 2.20M
0.965
Hexylmagnesium chloride
44767-62-6,109-99-9
Solution
C6H13ClMg
−25°C
Solubility in water: reacts
CCCCCC[Mg]Cl
144.93
144.93
AcroSeal™ Glass Bottle
0.9650g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only