missing translation for 'onlineSavingsMsg'
Learn More

Hexylene Glycol, Reagent, 98%, Spectrum™ Chemical

Catalog No. 18603129
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603129 500 mL Amber Glass Bottle
18603130 4 L Amber Glass Bottle
18603131 20 L Amber Glass Bottle
18603132 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603129 Supplier Spectrum Chemical Mfg Cor Supplier No. HE188500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

HE188, 107-41-5, C6H14O2

Spectrum™ Chemical Hexylene Glycol, Reagent

Chemical Identifiers

CAS 107-41-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
InChI Key SVTBMSDMJJWYQN-UHFFFAOYNA-N
IUPAC Name 2-methylpentane-2,4-diol
SMILES CC(O)CC(C)(C)O

Specifications

CAS Max % 100%
Boiling Point 197°C (386.6°F)
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Hexylene Glycol