missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Hexylene glycol

Catalog No. 11210018KG
Encompass
Change view
Click to view available options
Quantity:
18 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11210018KG 18 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11210018KG Supplier Sigma Aldrich Supplier No. 11210018KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexylene glycol

Specifications

CAS 107-41-5
Melting Point -40°C (lit.)
Density 0.925 g/mL (at 25°C (literature))
Boiling Point 197°C (lit.)
Molecular Formula C6H14O2
Refractive Index n20/D 1.427 (literature)
Linear Formula CH3CH(OH)CH2C(CH3)2 OH
Quantity 18 kg
MDL Number MFCD00004547
Synonym (+/-)-2-Methyl-2,4-pentanediol; MPD
Molecular Weight (g/mol) 118.17
Percent Purity 99%
Show More Show Less