missing translation for 'onlineSavingsMsg'
Learn More

Hexylcyclohexane 98.0+%, TCI America™

Catalog No. H013710ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H013710ML 10 mL
H013725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H013710ML Supplier TCI America Supplier No. H013710ML
Only null left

Chemical Identifiers

CAS 4292-75-5
Molecular Formula C12H24
Molecular Weight (g/mol) 168.32
MDL Number MFCD00019403
InChI Key QHWAQXOSHHKCFK-UHFFFAOYSA-N
Synonym 1-Cyclohexylhexane
PubChem CID 20283
IUPAC Name hexylcyclohexane
SMILES CCCCCCC1CCCCC1

Specifications

Color Colorless
Boiling Point 221°C
Quantity 10 mL
Formula Weight 168.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexylcyclohexane